Would you like to log in again?
SP Bel-Art H-B Durac® Calibrated Thermocouple Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Durac™
B60911-0200 B60901-1000 B60901-0800 B60901-1100
76486-050CS 1311.88 USD InStock
76486-050 76486-052 76486-046 76486-048
SP Bel-Art H-B Durac® Calibrated Thermocouple Thermometers, Bel-Art Products, a part of SP
Thermometers Digital Thermometers
Used for temperature measurement in life science research, kilns, gas turbine exhaust, diesel engines, and other industrial processes, H-B Calibrated Thermocouple Thermometers provide measurement throughout a broad temperature range and the ability to utilize multiple probes at one time.
  • Wide temperature range allows accurate monitoring at extreme temperatures
  • Max/min/average temperature recording capability
  • Data hold
  • Software CD (where included) for real time data collection and logging via USB
  • Bluetooth® app available for use with B60911-0200
Certifications: Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE-marked. Note: calibration applies to included probe and tested port only.
Ordering information: Probes for wider temperature ranges available (76486-054 and 76486-056) or use alternatively purchased probes; see chart for probe types accepted by each unit.
Delivery information: Includes two K-type beaded wire probes (–20 to +250 °C capability); one is calibrated and serialized.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty