Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NEB®10-beta Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs
NEB®
C3019H C3019P C3019I C3019P C3019H C3019I
76486-072EA 589 USD InStock
76486-072 CA76486-072 101417-920 200067-180 CA101417-920 CA200067-180
NEB®10-beta Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
NEB 10-beta Competent E. coli is a derivative of the popular DH10B. It is T1 phage resistant and endonuclease I (endA1) deficient for high-quality plasmid preparations.
  • High efficiency strain ideal for cloning large plasmids and BACs
  • Available in single-use vials, 200 µl vials, and electrocompetent format
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
  • Free of animal products
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty