Would you like to log in again?
COVERALL BA BASIC ZIP FRNT COLLR XL CS1
Supplier: PIP
Coverall, BA Microporous Basic, with collar, against dry particulates, liquid sprays without sacrificing breathability, Full length zipper front and strength for easy donning, ANSI Sized for wide range of motion, Elastic waistband for improved fit, Extremely low linting, Latex free, Size: xl
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVERALL BA BASIC ZIP FRNT COLLR XL CS1 76486-394 3600/XL Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty