Would you like to log in again?
APRON BA WHITE PP DISP 28X36IN 1SZ CS1
Supplier: PIP
Apron, BA White, 28x36in, Disposable, Base material: Polypropylene/Film Laminate/Polypropylene, Passes NFPA 99 Antistatic test, Low Lint, Passes U.S. ASTM 1670 standard for blood penetration, One Size fits most
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
APRON BA WHITE PP DISP 28X36IN 1SZ CS1 76486-478 3622 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty