Would you like to log in again?
Straight Sided Rounds, Clear, Tall, Wide Mouth
Supplier: Qorpak
GLC-05728 GLC-01655 GLC-05649 GLC-06845 GLC-05641 GLC-01654 GLA-00860 GLC-01658 GLC-01708 GLA-00866 GLC-07874 GLC-01656
16152-314CS 319.98 USD InStock
16152-314 16195-893 76239-640 36318-800 16152-318 76487-334 76239-642 16054-164 76487-332 16054-162 16054-196 36319-132
Straight Sided Rounds, Clear, Tall, Wide Mouth
Bottles Glass Bottles
Clear/flint straight sided round jars are designed without shoulders for maximum storage capacity and content visibility.
  • USP certified Type III (soda lime) glass
  • Cost-effective vacuum and ionization process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles from the container
  • Bottles cleaned and certified for semi-volatiles are pre-cleaned based on OSWER directives from the EPA
These multipurpose jars are very versatile. Perfect for pharmaceutical or general use, product storage, and field or plant sampling projects.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty