Would you like to log in again?
CELL FACTORY SYS 10-LAYER STD CLOSED CS2
Supplier: Thermo Fisher Scientific
Cell Factory System, Standard Closed, Material: Polystyrene, Height: 32.1 cm, Length: 47.2 cm, Temperature Range: 0C - 60C, Width: 39 cm, Disposable, Sterile, Culture Area: 6320cm2, Surface Treatment: Nunclon delta, Tray Layers: 10, Volume: Working: 2000 mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CELL FACTORY SYS 10-LAYER STD CLOSED CS2 76487-386 140472 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty