Would you like to log in again?
Sepharose™ Streptavidin High Performance Affinity Chromatography Media
Supplier: Cytiva
Sepharose™
90100484 90100484
76488-174EA 3259.05 USD InStock
76488-174 CA76488-174
Sepharose™ Streptavidin High Performance Affinity Chromatography Media
Protein Purification Protein Purification Resins
Streptavidin Sepharose™ High Performance is an affinity chromatography resin designed for fast and reliable, high resolution purification of biotinylated biomolecules.
Extremely useful for exploiting eith.er the strong interactions of biotin and streptavidin or the somewhat weaker interactions of 2-iminobiotin and streptavidin
34 µm bead size for improved resolution.
Purified Streptavidin isolated from Streptomyces avidinii is immobilised on Sepharose™ High Performance. The base matrix is a rigid, highly cross-linked beaded agarose with high chemical stability. The immobilised streptavidin binds biotin and biotinylated substances through affinity chromatography.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty