Would you like to log in again?
LiquidGuard™ Plus Sterile Cleanroom Coveralls, Apex Aseptic Products
Supplier: Apex
SGR4902PLAWHL SGR4902PLAWHXL SGR4902PLAWHS SGR4902PLAWHM SGR4902PLAWH3XL SGR4902PLAWH2XL SGR4902PLAWH4XL
76488-260CS 288.6 USD InStock
76488-260 76488-262 76488-264 76488-256 76488-266 76488-258 76488-268
LiquidGuard™ Plus Sterile Cleanroom Coveralls, Apex Aseptic Products
Disposable Garments
Sterile LiquidGuard™ Plus cleanroom coveralls feature a zipper front closure, without hood and booties.
  • Made with high strength LiquidGuard™ Plus microporous fabrics
  • Pass Blood Penetration Test per ASTM F1670
  • Pass ChemoDrug Permeation Test per ASTM F739-12
  • Anti-static treated, latex-free, TSE-free, and BSE-free
  • Easy breathability and comfortable to wear
  • Zipper front, no hood, no booties
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Gamma irradiated, validated sterile to SAL 10⁻⁶
  • Suitable to use in ISO Class 5 or higher cleanrooms
Inside seams are bonded with Teal color fabrics to indicate areas safe to touch inside the garment. Tunneled elastic cuffs with thumb loops help keep sleeves in place and prevent bare arms from exposure. Uniquely designed Clean-Don™ gowning system incorporates inside out folding and telescope leg method to ensure the wearer’s hands are not contaminating the outside of the sterile garments and the coverall’s legs are not touching the floor.
Each piece is individually packaged, triple bagged in plastic bags with easy tear notches, and labeled with item code, product name, lot number, production date, expiration date, and fixed with gamma irradiation indicator dot. Manufactured in a controlled environment.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected. Pass Blood Penetration Test per ASTM F1670. Pass ChomoDrug Permeation Test per ASTM F739-12.
Delivery information: Each case includes a Certificate of Conformance and Certificate of Sterility.
Packaging: Coverealls are packaged 1 piece per package, triple bagged, 25 pieces per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty