Would you like to log in again?
COVERALL W/HOOD BOOTIES WHITE 3X CS25
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Coverall, Sterile, LiquidGuard, attached hood, attached booties, pass blood penetration test, breathable, low particle shedding, rated category 1 per Helmke Drum Test, manufactured in controlled environment, Gamma Irradiated validated sterile to SAL 10-6, White, 3XL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVERALL W/HOOD BOOTIES WHITE 3X CS25 76488-304 S4903WH3XL Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty