Would you like to log in again?
Sterile Dupont® Tyvek™ Sleeve Covers, Apex Aseptic Products
Supplier: Apex
SGR1919TYWH18T-18-200 SGR1919TYWH21T-200 SGR1919TYWH21T-500
76488-322CS 558.4 USD InStock
76488-322 76488-324 76488-326
Sterile Dupont® Tyvek™ Sleeve Covers, Apex Aseptic Products
Disposable Garments
Sterile sleeve covers made with Dupont® Tyvek™ fabrics and are suitable for cleanroom use.
  • Dupont® Tyvek™ fabrics
  • Fluid resistant and breathable
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Latex-free, TSE-free, BSE-free
  • Manufactured in a controlled environment
  • Gamma irradiated validated sterile to SAL 10⁻⁶
  • Suitable to use in ISO Class 5 or higher cleanrooms
Sleeves feature attached thumb loops to help keep them in place. Tunneled sewing greatly reduces particle shedding. A special folding system helps to ensure proper aseptic donning.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected.
Delivery information: Each package includes a gamma irradiation indicator dot, and each shipping case is affixed with a Certificate of Conformance and Certificate of Sterility.
Packaging: Sleeves are packaged 2 pieces per package, triple bagged.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty