Would you like to log in again?
PEGuard Impervious Shoe Covers, Apex Aseptic Products
Supplier: Apex
2660A-BL/XL 2660A-B2XL 2660WH2XL 2660WHS/M 2660A-BS/M 2660WHL/XL
76488-360CS 83.3 USD InStock
76488-360 76488-350 76488-352 76488-354 76488-356 76488-358
PEGuard Impervious Shoe Covers, Apex Aseptic Products
Disposable Garments
PEGuard Impervious Shoe Covers are made of durable, skid-resistant PE coated non-woven fabrics. These garments are impervious and lint-free.
  • Durable, tear-resistant, water-resistant
  • Anti-skid treated to help prevent accidental falls
  • Latex-free, Silicone-free, and nonsterile
  • Low particle shedding, category 1 per Helmke Drum Test
  • Manufactured in controlled environment
  • Reasonable cost, affordable for everyday use
PEGuard Impervious Shoe Covers are made with specially treated, heavy-duty PE coated non-woven fabrics. They are ideal for use in pharmaceutical manufacturing facilities, animal labs, research centers, medical establishments, and cleanroom environments.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 certified. FDA registered and inspected.
Packaging: Shoe covers are packaged 50 per bag, 300 per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty