Would you like to log in again?
LAB COAT NO PKT LEVEL3 MED BLUE 2X CS100
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Lab coat, Extra-safe, Non-sterile, No pockets, Level 3, SMS fabrics, highly fluid resistant, knee length, snap front, knit cuffs, manufactured in controlled environment, low particle shedding, rated category 1 per Helmke drum testing, Medical Blue, 2XL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAB COAT NO PKT LEVEL3 MED BLUE 2X CS100 76488-446 NP3660MB2XL-100 Case of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty