CompletePAK® is a range of primary packaging components that are ready-to-use for most drug applications.
- Off-the-shelf, ready-to-use products
- One source for total packaging solutions
- Variety of different sterile components; a kit for any application
- Complete manufacturing traceability
- CompletePAK Amber for light-sensitive content packaging
- CompletePAK ModPAK bulk configurations
Each kit component is particulate cleaned, depyrogenated, and sterilized, arriving in the lab ready-to-use. Vials are packed in a sterile polypropylene tray which can be used as a filling aid. Kit packaging is sized to compliment manual aseptic filling processes.
The CompletePAK® “single source” focus can reduce customer’s supply chain and time to market. Vials, stoppers, and seals are particulate cleaned, depyrogenated and sterilized, arriving in the lab ready-to-use.
Certifications: 1. USP <788> particulate Matter in Injections
2. USP <85> Bacterial Endotoxin Test
3. Lot certified Sterile to SAL <10-6
Delivery information: Batch-specific Certificates of Analysis showing compliance with relevant USP standards are included in each kit.