Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Interconnect Cord, with 2 Banana Plugs, Transforming Technologies
Supplier: Transforming Technologies
CC3000NR
76489-586EA 5.46 USD InStock
76489-586
Interconnect Cord, with 2 Banana Plugs, Transforming Technologies
Grounders Grounding Wires
Two standard banana plugs allow the cord to connect two devices such as mats or ground cords that feature banana jacks.
  • Use to connect two items together
  • Each end terminates with a standard banana plug
  • Length is 8'
  • No resistor
The Interconnect Cord features 2.5 mm polyurethane insulation that offers excellent cord life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty