Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Dual Conductor Coil Cords, Transforming Technologies
Supplier: Transforming Technologies
CC3050R CC3100 CC3100R CC3200 CC3200R CC3050
76489-588EA 24.24 USD InStock
76489-588 76489-598 76489-596 76489-594 76489-592 76489-590
Dual Conductor Coil Cords, Transforming Technologies
Grounders Grounding Wires
A double insulated jacket provides incredible durability and a wide diameter straight plug makes it easy to insert and remove the cord from a remote input jack.
  • If one conductor is damaged, the operator will maintain the path-to-ground through the other conductor
  • 3.4 mm straight mono plug
  • 1 meg ohm series resistor
  • 5, 10, and 20' lengths
  • Right angle plug models are also available
Used in conjunction with Dual Conductor Constant Workstation Monitors, these Dual Conductor Coil Cords provide unmatched reliability and value. A double insulated jacket provides incredible durability and a wide diameter straight plug makes it easy to insert and remove the cord from a remote input jack. Cords come standard in 5, 10 or 12' lengths.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty