Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Stud Connection, 10 mm to 10 mm, Transforming Technologies

Supplier: Transforming Technologies
CS0128
76489-636PK 14.67 USD InStock
76489-636
Stud Connection, 10 mm to 10 mm, Transforming Technologies
Grounders Grounding Wires
This Snap Adapter Set is compatible and interchangeable with all major snap manufactures.

  • Steel adapter has a double 10mm socket construction
  • Used to convert 10 mm snaps, primarily with anti-static matting and ground cords
  • Snap adapter connects a 10 mm stud to a 10 mm stud

Also available in a socket-to-socket connection with the CS0128-socket.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty