Would you like to log in again?
Conductive Rubber V-Groove Floor Mats, Black, Transforming Technologies
Supplier: Transforming Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FM104872 FM103X32 FM102448 FM103648 FM104X32
76489-654EA 297.41 USD InStock
76489-654 76489-652 76489-660 76489-658 76489-656
Conductive Rubber V-Groove Floor Mats, Black, Transforming Technologies
Floor Mats Permanent Floor Mats
This conductive ESD floor runner has a non-slip, grooved surface that provides excellent traction and allows the mat to be easily cleaned.
  • Conductive ESD floor runner
  • V-groove
  • Gray
  • ⅛" thick
The ribbed matting is also very resilient, allowing it to stand up to heavy foot traffic while still maintaining its ESD protection. The material also offers moderate chemical resistance. The matting is available in 4×32' rolls and an assortment of lengths. The Transforming Technologies GrooveSTAT™ Conductive Rubber Matting provides the electrical properties specified by ESDA standards for floor mats. Most common ESD runners available are made with dissipative vinyl, which does not comply with ESD standards.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty