Would you like to log in again?
Mat Interconnect Cord, with Two 10 mm Male Snaps, Transforming Technologies
Supplier: Transforming Technologies
FM2525NR
76489-684EA 11.49 USD InStock
76489-684
Mat Interconnect Cord, with Two 10 mm Male Snaps, Transforming Technologies
Grounders Grounding Wires
The ground cord is used to interconnect two monitored work surfaces when unable to connect individually to ground.
  • 10' length
  • No resistor (zero meg)
  • 10 mm male snaps
Straight cord with male snap fasteners at each end and no resistor. It is used to interconnect two monitored work surfaces together. Supplied with two molded low profile mushroom caps with ³/₈" (10 mm) male snaps to ensure a secure connection to mats.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty