Would you like to log in again?
Dissipative V-Groove Vinyl ESD Floor Mats, Transforming Technologies
Supplier: Transforming Technologies
FM348720GY FM34872GY FM33672GY FM336720GY
76489-688EA 491.92 USD InStock
76489-688 76489-686 76489-692 76489-690
Dissipative V-Groove Vinyl ESD Floor Mats, Transforming Technologies
Floor Mats Permanent Floor Mats
Designed to provide a static-safe work area for a wide variety of applications where ESD properties are required.
  • Dissipative ESD floor runner
  • V-groove
  • Gray
V-groove floor runner mats can reduce slips and falls by increasing traction, even when exposed to moisture and chemicals. Transforming Technologies’ VinylStat FM3 V-Groove solid floor runners are made from a homogeneous polymer mix of durable PVC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty