Would you like to log in again?
Table Mat Ground Kit, Common Point Ground and Universal Snap Kit, Transforming Technologies
Supplier: Transforming Technologies
GK1000
76489-714EA 13.2 USD InStock
76489-714
Table Mat Ground Kit, Common Point Ground and Universal Snap Kit, Transforming Technologies
Grounders Grounding Wires
Easy to install kit for grounding ESD table mats.
  • Includes 15' low profile ground cord, and a universal snap
  • Universal snap kit easily installs a ground snap into a range of mat thicknesses
  • Includes two screws for thick and thin materials
The CP2522 is a low profile common point ground with two banana jacks which allow two workers the ability to plug into one ground point. The molded connection point houses a 1 meg Ohm, ¹/₄ watt resistor.
The ground cord attaches to table mats via the Universal Snap Kit. The Universal Snap Kit includes a 10 mm stud, 10 mm socket, threaded backing washer, conical nut, long screw, short screw, and ESD common point ground label. Everything needed to install a stud or socket on all soft materials up to 0.375" thick. Screws are 6-32 flat head Phillips machine screws; one 0.25" long and one 0.375" long. Punch a hole through the mat and screw on the socket or stud, whichever is preferred. All that is needed is a Phillips screwdriver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty