Would you like to log in again?
Heel Grounder, with 1.25" Cup and Hook and Loop Closure, Transforming Technologies
Supplier: Transforming Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HG1360R HG1360B HG1370
76489-720EA 7.2 USD InStock
76489-720 76489-718 76489-722
Heel Grounder, with 1.25" Cup and Hook and Loop Closure, Transforming Technologies
Grounders Heel and Toe Grounders
Heel Grounders are constructed from durable materials and feature a stretch hook and loop closure strap to maximize fit and comfort.
  • Ground mobile personnel with long-lasting stretch hook and loop ESD heel grounders
  • Reversible cup design allows for longer life
  • When one side becomes worn, flip the strap and use the other side to extend the life
  • A buried 1 meg resistor is built into the strap for safety and to comply with ANSI ESD S2020
Heel grounders connect the person wearing them to ground via a proper floor mat or flooring material. Wearing the conductive ribbon inside the shoe or sock assures proper electrical contact with the user. The ribbon connects to the conductive rubber to complete the circuit between the operator and the static controlled floor or mat.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty