Would you like to log in again?
Hook and Loop Heel Grounders, 1 Meg, Transforming Technologies
Supplier: Transforming Technologies
HG1910Y HG1910B
76489-730EA 6.58 USD InStock
76489-730 76489-728
Hook and Loop Heel Grounders, 1 Meg, Transforming Technologies
Grounders Heel and Toe Grounders
Heel grounders connect the wearer to ground via a proper floor mat or flooring material.
  • Reversible cup design for long life
  • Stretch hook and loop closure strap allows maximum fit and comfort
  • Available in blue and yellow
The HG1910 series Heel Grounders are constructed from durable materials and feature a stretch hook and loop closure to maximize fit and comfort. The 1" reversible cup is designed to extend the life of the device.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty