Would you like to log in again?
Sole Grounders, 2 Meg, Transforming Technologies
Supplier: Transforming Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HG6052 HG6012 HG6022 HG6032
76489-740EA 15.54 USD InStock
76489-740 76489-738 76489-736 76489-734
Sole Grounders, 2 Meg, Transforming Technologies
Grounders Heel and Toe Grounders
Full coverage sole grounders provide 20 times the surface contact of heel grounders.
  • Wider and more consistent contact area ensures grounding
  • Rapid and complete static dissipation due to constant contact through entire walking motion
ESD Sole Grounders (or ESD Foot Grounders) offer over 20 times the surface contact of standard ESD heel straps. This surface area advantage extends the life of the sole grounder over standard heel grounders, which fail over time due to even the slightest contamination, necessitating replacement or cleaning, wasting time, and costing money.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty