The 9010 Series ESD jackets provide high-quality ESD protection at a great value.
- Level 3 ESD jacket
- Maintains consistent continuity after 100 wash cycles
- Snap-front closure
- Three patch pockets
The 9010 Series jackets are fabricated from lightweight 90% polyester and 10% carbon fabric for unmatched operator comfort. Jackets feature ESD knit cuffs, and lapel style collar.
Available in blue, black, white, teal, and maroon.