Would you like to log in again?
WB1600 Series Wrist Band and Coil Cord Set, Transforming Technologies
Supplier: Transforming Technologies
WB1643
76490-244EA 9.47 USD InStock
76490-244
WB1600 Series Wrist Band and Coil Cord Set, Transforming Technologies
Grounders Wrist Grounders
The WB1600 Series Wrist Band and Coil Cord set is the most versatile and cost-effective band set offered by Transforming Technologies.
  • Wrist strap is a durable and long-lasting method of grounding personnel
  • Made with a soft woven fabric that is comfortable to wear for long periods
  • Fabric band uses hypoallergenic silver-plated nylon thread for conductivity
  • Band will not irritate skin
The soft, blue woven fabric and 360° curved buckle of the wrist band allows the skin to breathe and makes it very comfortable to wear. The band uses hypoallergenic silver-plated nylon thread for conductivity and has a maximum resistance of 160 ohms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty