Would you like to log in again?
HOSE TRANSFER SS CRYOGNC 6FT LN2 SYSTEM
Supplier: PHC Corporation
Transfer hose, Cryogenic stainless steel, for LN2 system only. Must be Pre-installed, 6ft
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HOSE TRANSFER SS CRYOGNC 6FT LN2 SYSTEM 76490-338 CVKLN2H6 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty