Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
MXP Data Loggers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
MXPD-M MXP-M MXP-S MXPD-S MXP-RAO
76490-424EA 95.03 USD InStock
76490-424 76490-426 76490-420 76490-422 76490-428
MXP Data Loggers, Cryopak Verification Technologies
Data Loggers
The MXP data logger offers flexibility and longevity with a light sensor to detect and log if package is opened.
  • Long shelf life
  • Event logging feature
  • Internal digital temperature sensor
With its USB connectivity, the MXP data logger provides detailed reports in both PDF and Excel format, and is user friendly for both Windows and smartphone app use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty