Would you like to log in again?
SLIDE MICROSCOPE AUTOFROST 1MM CS1440
Supplier: CANCER DIAGNOSTICS, INC.
Microscope Slide, AutoFrost, 1mm Thickness, Non-Charged Clipped Corner, Painted surface suitable for printers, labeling and writing instruments. 90 degree (square) corner, White, Size: 75 x 25mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SLIDE MICROSCOPE AUTOFROST 1MM CS1440 76490-494 AF45-0 Case of 1,440 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty