Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Borosil® Reusable Class A Mohr (Measuring) Pipettes (Pipettes) with Individual Serial Numbers, Foxx Life Sciences®
Supplier: Foxx Life Sciences
BOROSIL®
7059P11D 7059P55A 7059P06A 7059P09D 7059P11A 7059P41D 7059P02A 7059P05D 7059P55D 7059P02D 7059P01D 7059P05A 7059P42A 7059P42D 7059P06D 7059P22D 7059P22A 7059P01A 7059P09A 7059P41A
76491-340CS 155.45 USD InStock
76491-340 76491-344 76491-322 76491-310 76491-332 76491-320 76491-342 76491-330 76491-326 76491-314 76491-336 76491-346 76491-324 76491-312 76491-334 76491-308 76491-318 76491-328 76491-338 76491-316
Borosil® Reusable Class A Mohr (Measuring) Pipettes (Pipettes) with Individual Serial Numbers, Foxx Life Sciences®
Pipettes Measuring (Mohr) Pipettes
These Borosil brand pipettes are made of specially-selected heavy wall machine-drawn borosilicate accurate bore glass tubing.
  • Markings are made in permanent amber stain diffused into the surface of the glass for better visibility and durability
  • Graduated for delivery from zero mark to the last graduation mark
They have precision formed jets, and bevel ground ends that resist chipping. The graduation marks are made in durable white enamel or amber stain, resistant to normal laboratory reagents and repeated washing. All the Pipettes are individually measured, and then marked. The uniformity of bore ensures accuracy between any two readings. The Pipettes can be autoclaved and, if necessary, can be flamed for sterilization.
Certifications: Complies with IS, ISO / DIN 835.
Delivery information: Supplied with individual or bulk calibration certificate showing actual volume dispensed and providing traceability to national standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty