Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Borosil® Reusable Class B Mohr (Measuring) Pipettes (Pipettes), Foxx Life Sciences®
Supplier: Foxx Life Sciences
BOROSIL®
7060P22D 7060P42D 7060P55D 7060P41D 7060P09D 7060P02D 7060P05D 7060P11D 7060P01D 7060P06D
76491-366CS 224.25 USD InStock
76491-366 76491-354 76491-364 76491-352 76491-348 76491-358 76491-356 76491-362 76491-350 76491-360
Borosil® Reusable Class B Mohr (Measuring) Pipettes (Pipettes), Foxx Life Sciences®
Pipettes Measuring (Mohr) Pipettes
These Borosil brand pipettes are made of specially-selected heavy wall machine-drawn borosilicate accurate bore glass tubing.
  • Markings are made in permanent amber stain diffused into the surface of the glass for better visibility and durability
  • Graduated for delivery from zero mark to the last graduation mark
They have precision formed jets, and bevel ground ends that resist chipping. The graduation marks are made in durable white enamel or amber stain, resistant to normal laboratory reagents and repeated washing. All the Pipettes are individually measured, and then marked. The uniformity of bore ensures accuracy between any two readings. The Pipettes can be autoclaved and, if necessary, can be flamed for sterilization.
Certifications: Complies with IS, ISO / DIN 835.
Delivery information: Supplied with individual or bulk calibration certificate showing actual volume dispensed and providing traceability to national standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty