Would you like to log in again?
Borosil® Reusable Test Tubes without Rim, Foxx Life Sciences
Supplier: Foxx Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Borosil®
9820U02 9820U04 9820U16 9820U18 9820U12 9820U17 9820U09 9820U03 9820U05 9820U06 9820U08 9820U19 9820U01 9820U10 9820U11 9820U07
76491-542CS 172.17 USD InStock
76491-542 76491-552 76491-530 76491-540 76491-550 76491-546 76491-534 76491-556 76491-544 76491-532 76491-554 76491-528 76491-538 76491-548 76491-526 76491-536
Borosil® Reusable Test Tubes without Rim, Foxx Life Sciences
Tubes Test Tubes
Test tubes constructed of borosilicate 3.3 glass, without rim.
  • Excellent chemical resistance
  • Thick walled, mechanically resistant
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Designed without a rim
  • Graduation range: Type I
The large opening can even accommodate large volume funnels, giving easy access to scoops or spatulas.
Amber to protect light-sensitive materials.
Certifications: In compliance with ISO/DIN 4142 standards.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty