Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Environmentally Friendly Thermometers, Easy Read
Supplier: VWR
VWR®
60300-0111 60303-0011 60304-0211 60301-0211 60302-0011 60301-0311
76491-596CS 470.19 USD InStock
76491-596 76491-600 76491-598 76491-604 76491-602 76491-606
VWR® Environmentally Friendly Thermometers, Easy Read
Thermometers Analog Thermometers
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • Enviro safe liquid
  • Eliminates reading errors
  • Non-toxic, non-hazardous, biodegradable liquid
Includes multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability, non roll fitting and individual serial number.
Certifications: Traceable NIST. Envirokleen certified by ChemTel (an independent laboratory).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty