Would you like to log in again?
VWR® Plus Dry Block/Incubator Liquid-in-Glass Thermometer, Organic Liquid Fill
Supplier: VWR
VWR®
60701-0111
76491-624CS 667.73 USD InStock
76491-624
VWR® Plus Dry Block/Incubator Liquid-in-Glass Thermometer, Organic Liquid Fill
Thermometers Analog Thermometers
Contains blue organic liquid against white back glass.
  • Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team
Specifically for heating blocks and dry baths.
Certifications: Traceable to NIST.
Delivery information: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy indicating accuracy traceable to NIST.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty