Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Liquid-in-Glass Oven Thermometers
Supplier: VWR
VWR®
60803-0611 60803-1011
76491-640CS 220.07 USD InStock
76491-640 76491-638
VWR® Liquid-in-Glass Oven Thermometers
Thermometers Analog Thermometers
These oven thermometers contain red organic liquid against white back glass.
  • Partial immersion thermometers feature a round glass bulge in the body to hold them in place
  • PTFE coating helps prevent liquid and glass contamination if the thermometer breaks
  • Individually serialised
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty