Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Dry Block/Incubator Liquid-in-Glass Thermometer
Supplier: VWR
VWR®
60810-1511
76491-644CS 308.47 USD InStock
76491-644
VWR® Dry Block/Incubator Liquid-in-Glass Thermometer
Thermometers Analog Thermometers
Contains red organic liquid against white back glass.
  • For use in all standard heating blocks and incubators
  • PFA safety coating to keep liquid and glass encapsulated in the event of breakage
  • Individually serialized
Temperature range specific for Incubators and dry blocks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty