Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Pocket Thermometers
Supplier: VWR
VWR®
608701811 60870-1911 60870-0911 60870-1811 60870-2111 60870-1611 60870-1211 60870-1711 60870-2511 60870-3811 608702511 608702611 60870-2611 60870-1111 608701611 608700911
89095-550CS 114.74 USD InStock
89095-550 CA89095-538 89095-538 76491-650 89095-554 89095-532 CA89095-542 76491-648 89095-531 89095-542 CA89095-550 76491-646 89095-536 89095-534 CA89095-532 CA89095-554
VWR® Pocket Thermometers
Thermometers Analog Thermometers
Pocket glass thermometers feature permanently fused markings.
  • Red spirit-filled
  • Includes a heavy-duty case
  • Compact size
  • Glass fabrication
  • Permanently fused markings
Markings are easy to read against thermometer background.
Order Now
ORDER
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty