Would you like to log in again?
VWR® Thermometer Case for Liquid-in-Glass Thermometers
Supplier: VWR
VWR®
61400-9111
76491-684CS 65.07 USD InStock
76491-684
VWR® Thermometer Case for Liquid-in-Glass Thermometers
Thermometer Holders
Open cases feature a screw cap with ring and small openings around bulb for circulation.
  • Screw cap
  • Ring for hanging
  • Metal or nickel-plated brass
  • Lower section surrounding the bulb with small openings for circulation
This case protects your thermometers from accidental breakage storage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty