Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Single Channel Electronic Timer with Triple Alarms and Certificate of Calibration
Supplier: VWR
VWR®
61700-3611
76491-690CS 499.86 USD InStock
76491-690
VWR® Single Channel Electronic Timer with Triple Alarms and Certificate of Calibration
Timers Digital Timers
Choice of vibration, sound, and/or light alarms make this timer ideal for use in high noise locations.
  • Timing capacity: 99 minutes, 59 seconds
  • Count direction: up/down
  • Clock function
Vibration, light, and loud buzzer alarms can function at one time, or be independently shut off.
Certifications: Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert. #2448.01), against equipment whose calibration is traceable to NIST. CE marked and RoHS compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty