Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® 4-Channel Switchable Electronic Timer
Supplier: Avantor
VWR®
61700-3711
76491-692CS 499.86 USD InStock
76491-692
VWR® 4-Channel Switchable Electronic Timer
Timers Digital Timers
Timer features a built-in a whiteboard with marker and eraser, which helps to keep track of time and other critical information.
  • Switchable count up and down with elapsed time displayed after countdown
  • Switchable capacity of 23 hours, 59 minutes to 59 minutes, 59 seconds
  • Audible alarm for each channel
  • Last time set memory
  • Audible confirmation for a data entry key press
Mounting options: lay flat on bench, flip-open stand for vertical view, and a magnetic back for metal surface attachment.
Certifications: Individually tested and inspected using NIST in A2LA accredited laboratory (certification #2448.01), assuring conformance to ISO 17025:2005. RoHS compliant.
Delivery information: Supplied with batteries and certificate of calibration. Requires two AAA batteries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty