Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Digital Stopwatch, with Clock and Timer
Supplier: VWR
VWR®
61700-5211
76491-694CS 1316.71 USD InStock
76491-694
VWR® Digital Stopwatch, with Clock and Timer
Timers Digital Stopwatches
This electronic stopwatch includes a calendar and a clock.
  • Three-line display: time, countdown, and pacer
  • Timing capacity: 10 hours
  • 1/100 second timing
  • Stores 30 lap times and calculates lap time average; allows for 99 split lap times
  • 10/320 second pacer
Supplied with multi-language instructions (English, French, German, Italian, Portuguese, and Spanish).
Certifications: Tested and calibrated in a triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: Supplied with one CR2032 battery, certificate of calibration, individual serial number, and multi-language (English, French, German, Italian, Portuguese, and Spanish) instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty