Would you like to log in again?
VWR® Sleeves, Made in USA
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
353 353TL
76491-714CS 363.56 USD InStock
76491-714 76491-708
VWR® Sleeves, Made in USA
Disposable Garments
VWR® Made in USA Sleeves are an ideal option for single-use needs. Made with the specially formulated Alliance breathable microporous fabric.
  • Individually bagged
  • Available with thumbloop configuration
  • Made with Alliance breathable microporous fabric
  • Made in USA
A key part of the VWR® Made in USA PPE line, these cleanroom sleeves are 21" in length to provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
The Alliance fabric is designed to lower particulate levels by providing extremely smooth outer fabric surface that lowers particle cling and build-up in cleanroom applications. Alliance products have a unique antistatic system that assures static dissipation in sensitive cleanroom applications, while providing a liquid-proof barrier that ensures protection from hazardous substances used in Biotech and Pharmaceutical cleanrooms.
Packaging: Bagged 1 pair per bag, 100 pairs per case. Packed in poly-lined bag in brown corrugated box.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty