Would you like to log in again?
InocuLoop Inoculating Loops, Caplugs
Supplier: Caplugs
333-5013-G1I 333-5010-Y10 333-5002-B1I 333-5001-B10 333-5012-G10 333-5011-Y1I
76492-640CT 86.03 USD InStock
76492-640 76492-642 76492-644 76492-646 76492-636 76492-638
InocuLoop Inoculating Loops, Caplugs
Spreaders Inoculating Loops and Needles
Inoculoop disposable inoculating loops have patented four-sided spreaders, enabling the inoculation of a petri dish with one, not multiple, disposable loops. Make bacterial inoculation and quantitative procedures faster and more efficient.
  • Made of flexible plastic for easy spreading
  • Can also be used as a picker for isolating colonies
  • Pre-sterilized Inoculoops are gamma-irritated
Suitable in anaerobic chambers, in the field and even in satellite metal loops.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty