Would you like to log in again?
KIT LOW DISPERSION PIPING
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Low Dispersion Piping Kit, Semi-Micro Mixer, Sample Loop(40Ul), As-Column Pipe(0.1Mmi.D.Sus)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT LOW DISPERSION PIPING 76493-186 892-0821 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty