Would you like to log in again?
Test Tubes, Caplugs
Supplier: Caplugs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
214-2023-010 214-2009-010 214-2367-B30 214-2345-03N 214-2355-030 214-2070-010 214-2309-030 214-2030-010 214-2012-010 214-2003-010 214-2065-01N 214-2088-G50 214-2002-010 214-2367-Y30 214-2001-010 214-2367-R30 214-2068-G50 214-2131-010 214-2097-010 214-2380-030 214-2367-030 214-2079-G60 214-2313-030 214-2099-G50
76493-638BG 58 USD InStock
76493-638 76493-648 76493-658 76493-668 76493-646 76493-678 76493-656 76493-644 76493-666 76493-654 76493-676 76493-642 76493-664 76493-674 76493-652 76493-640 76493-662 76493-684 76493-672 76493-650 76493-660 76493-682 76493-670 76493-680
Test Tubes, Caplugs
Tubes Test Tubes
Test tubes provide clarity and can be used for mild acidic or alkali solutions, alcohol and salt, with exceptions of oxidizing acids and high concentrate aclohols.
  • Provided with or without colors
  • Tubes are bulk packed, non-sterile
  • Polystyrene tubes withstand up to 8200×g
  • Polypropylene tubes withstand up to 10000×g
  • 12×75 mm tubes are available with internally ribs for use in antibody coating and/or protein adsorptions, six slanted ribs increase useful surface area
The polypropylene tubes are chemically resistant and are appropriate for use with organic solvents and stronger acids, alkalis, alcohols, and chlorinated and aromatic hydrocarbons.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty