Polyflow® cartridges are optimized for use in many applications. They feature a random-fiber polypropylene depth matrix that provides excellent retention effciencies and onstream life. The unique calendering process produces depth media with an absolute rating and superior dirt-holding capacity.
- High-retention depth matrix
- High flow rate
- Wide variety of configurations and ratings
- Economical prefiltration
These cartridges are thermally bonded from 100% virgin polypropylene to ensure a high level of cleanliness and chemical compatibility.
Certifications: In compliance with DIN ISO 4796-1 standards.
Delivery information: Supplied with screw cap and pouring ring.
Packaging: The bottles are packaged in shrinked-wrapped modules.