Would you like to log in again?
CARTRIDGE PP 226/FIN 30IN 3UM EPDM 5 MM
Supplier: PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: 226/Fin, Nominal Length: 30in, Filter Rating: 3um, Gasket O-ring: EPDM, Thickness: 0.200in (5 mm), Depth media: Pleated Polypropylene, provides excellent retention efficiencies and onstream life, optimized for use in liquid applications
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE PP 226/FIN 30IN 3UM EPDM 5 MM 76494-542 28-19730-030-11 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty