Would you like to log in again?
CARTRIDGE PP 226/FIN 5IN 0.2UM EPDM
Supplier: PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: 226/Fin, Nominal Length: 5in, Filter Rating: 0.2um, O-Ring: EPDM, Depth media: Pleated Polypropylene, provides excellent retention efficiencies and onstream life, optimized for use in liquid applications
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE PP 226/FIN 5IN 0.2UM EPDM 76494-804 25-10705-002-1-B Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty