Would you like to log in again?
CARTRIDGE 10IN 0.1UM FLAT TOP/222 SIL
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 10in, 0.1um Pleated PTFE membrane, Endcap: Flat Top / 222, Pharmaceutical variant, Silicone o-rings, manufactured entirely from fluoropolymers making them extremely resistant to a wide range of aggressive chemicals.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 10IN 0.1UM FLAT TOP/222 SIL 76495-450 ZCTP1-010E-PS Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty