Would you like to log in again?
CARTRIDGE 20IN 5U PP BF/226 BAYONT SILCN
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, 5.0um, Pleated Polypropylene media, End cap: BF / 226 Bayonet fitting, designed to guarantee removal of particulate from gas streams to protect sterilizing grade filters in pressurized systems.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 20IN 5U PP BF/226 BAYONT SILCN 76495-552 ZCPH2-005C Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty