Would you like to log in again?
CARTRIDGE 20IN 0.1UM PTFE MEMB 222/FLAT
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, .1um Pleated PTFE membrane, 222/Flat end cap with external EPDM o-rings, particularly suitable for sterilization and particulate removal from aggressive chemicals within a wide range of critical processing industries.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE 20IN 0.1UM PTFE MEMB 222/FLAT 76495-758 ZCMT2-010E-LE Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty